Specifications
- Antibody Type:Primary
- Antigen Symbol:SLC35D1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75931-986
- Supplier no.:600-401-EP3
Specifications
About this item
The solute carrier family Slc35 consists of at least 17 proteins that act as nucleotide sugar transporters localized to the Golgi apparatus and endoplasmic reticulum. The role of the ER-resident Slc family member Slc35D1 is to transport both UDP-glucuronic acid and UDP-N-acetylgalactosamine. These molecules can serve as substrates for chondroitin sulfate biosynthesis and mice lacking the Slc35D1 gene developed a lethal form of skeletal dysplasia with severe shortening of limbs and facial structures. Examination of epiphyseal cartilage in these mice revealed a decreased proliferating zone with round chrondrocytes, scarce matrices, and reduced proteoglycan aggregates. Loss of function mutations in human Slc35D1 cause Schneckenbecken dysplasia, a severe skeletal dysplasia.
Anti-Slc35D1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 39 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: SLC35D1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: