To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:SLC1A7
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75935-382
- Supplier no.:600-401-EN8
Specifications
About this item
The solute carrier family 1 member 7 (SLC1A7) protein was initially identified as a glutamate transporter coupled to a chloride conductance that is expressed primarily in the retina (1). More recent evidence indicates that is it widely expressed in peripheral tissues, also (2). Experiments have shown that SLC1A7 functions as a low-affinity/low-capacity glutamate transport, with an anion channel optimized for anion conductance in the negative voltage range (3). In the retina, SLC1A7 co-localizes with the serum and glucocorticoid dependent kinase SGK1, and its cell surface expression and activity is regulated SGK1 and its isoform SGK3 (4).
Anti-SLC1A7 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 61 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: SLC1A7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: