Specifications
- Antibody Type:Primary
- Antigen Symbol:SIGIRR
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75931-876
- Supplier no.:600-401-EK1
Specifications
About this item
SIGIRR is a member of the Toll-like receptor-interleukin 1 receptor superfamily. Members of this family are defined by the presence of an intracellular Toll-IL-1R (TIR) domain. The Toll-like receptors (TLRs) are signaling molecules that recognize different microbial products during infection and serve as an important link between the innate and adaptive immune responses. SIGIRR was originally identified through database analysis and was shown to have only one Ig domain as opposed to the normal three Ig folds seen in the TIR family. Similar to ST2, another TIR family member, it has been shown to negatively regulate IL-1 receptor and Toll-like receptor signaling. However, SIGIRR inhibits TLR-IL-1R signaling by dimerizing with TLR4, TLR5, TLR9, and IL-1R. It also associates with the down-stream TLR signaling proteins IRAK and TRAF6 in an IL-1-dependent fashion.
Anti-SIGIRR Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 46 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: SIGIRR
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: