Order Entry
United States
ContactUsLinkComponent
Anti-Protocadherin-Gamma-A1 Mouse Monoclonal Antibody (BAC) [Clone: S159-5]
Anti-Protocadherin-Gamma-A1 Mouse Monoclonal Antibody (BAC) [Clone: S159-5]
Catalog # 75931-220
Anti-Protocadherin-Gamma-A1 Mouse Monoclonal Antibody (BAC) [Clone: S159-5]
Catalog # 75931-220
Supplier Number:  200-306-W68
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Protocadherin-Gamma-A1
  • Clonality:
    Monoclonal
  • Clone:
    S159-5
  • Conjugation:
    Biotin
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Environmentally Preferable:
  • Storage Temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping Temperature:
    Wet Ice
  • Purification:
    Protein G purified
  • Cat. No.:
    75931-220
  • Supplier no.:
    200-306-W68

Specifications

About this item

The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes.
The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. Anti-Protocadherin Gamma (pan) is ideal for research in Neuroscience, Cell Adhesion and Cell Signaling.

Anti-Protocadherin Gamma (pan) Biotin Conjugated Antibody is suitable for Western Blots and Immunocytochemistry. Expect a band approximately ~100 kDa on specific lysates or tissues. It will cross react with all Gamma-protocadherins -A, -B, and -C. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Protocadherin-Gamma-A1
Clonality: Monoclonal
Clone: S159-5
Conjugation: BAC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: