To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antibody Type:Primary
- Antigen Name:Protocadherin-Gamma-A1
- Clonality:Monoclonal
- Clone:S159-5
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:
- Western Blot:Yes
- Size:100 µg
- Environmentally Preferable:
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Wet Ice
- Purification:Protein G purified
- Cat. No.:75931-222
- Supplier no.:200-302-W68
The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes.
The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. Anti-Protocadherin Gamma (pan) is ideal for research in Neuroscience, Cell Adhesion and Cell Signaling.
Anti-Protocadherin Gamma (pan) FITC Conjugated Antibody is suitable for Western Blots and Immunocytochemistry. Expect a band approximately ~100 kDa on specific lysates or tissues. It will cross react with all Gamma-protocadherins -A, -B, and -C. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: Protocadherin-Gamma-A1
Clonality: Monoclonal
Clone: S159-5
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: