Specifications
- Antibody Type:Primary
- Antigen Symbol:PLG
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Ambient
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
- Cat. No.:75931-056
- Supplier no.:200106208-0100
Specifications
About this item
Anti-Plasminogen antibody recognizes plasminogen, a precursor of plasmin. Plasmin is a serine protease that degrades fibrin clots and involved in embryonic development, tissue remodeling, tumor invasion, and inflammation. In addition to fibrin, Plasmin can also cleave fibronectin, thrombospondin, laminin, and von Willebrand factor. Plasmin can activate other zymogens such as tissue plasminogen activator, urokinase plasminogen activator, kallikrein, and factor XII. Deficiency in plasminogen increases susceptibility to thrombosis.
Anti-Plasminogen antibody has been assayed against 1.0 ug of Plasminogen in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature. A working dilution of 1:4,000 to 1:20,000 of the reconstitution concentration is suggested for this product.
Type: Primary
Antigen: PLG
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity: