Specifications
- Antibody Type:Primary
- Antigen Symbol:PDCL
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75930-762
- Supplier no.:600-401-DK4
Specifications
About this item
Phosducin-like proteins (PhLPs) are a conserved family of proteins with thioredoxin-like domains that were initially identified as modulators of G protein signaling (1,2). PDCL is widely expressed in normal human tissues and may participate directly in the regulation of calcium-evoked exocytosis in adrenal medullary chromaffin cells (3). It is an ethanol-responsive modulator of heterotrimeric G proteins (4).
Anti-PDCL Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 34 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: PDCL
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: