Order Entry
Puerto Rico
ContactUsLinkComponent
 
Anti-ORAI1 Mouse Monoclonal Antibody [Clone: [6D11A11]]
Catalog # 75930-638
undefined
Anti-ORAI1 Mouse Monoclonal Antibody [Clone: [6D11A11]]
Catalog # 75930-638
Supplier Number:  200-301-DH1

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    ORAI1
  • Clonality:
    Monoclonal
  • Clone:
    [6D11A11]
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2b
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Storage Temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping Temperature:
    Dry Ice
  • Purification:
    Protein A purified
  • Cat. No.:
    75930-638
  • Supplier no.:
    200-301-DH1

Specifications

About this item

Antigen stimulation of immune cells triggers Ca++ entry through Ca++ release-activated Ca++ (CRAC) channels. ORAI1 is a recently identified four-transmembrane spanning protein that is an essential component of CRAC. A missense mutation in this protein in humans is the cause of one fo rm of hereditary severe combined immune deficiency (SCID) which results in ablated T-cell Ca++ entry. It has been suggested that ORAI1 functions as a highly selective Ca++ plasma membrane channel that is gated through interactions with STIM1, the store-activated endoplasmic reticulum Ca++ sensor. ORAI1 often migrates at a higher than expected molecular weight in SDS-PAGE. This antibody is predicted to have no cross-reactivity to ORAI2 or ORAI3.

Anti-ORAI1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 33 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: ORAI1
Clonality: Monoclonal
Clone: [6D11A11]
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: