- Antibody Type:Primary
- Antigen Name:Neuroligin-1
- Clonality:Monoclonal
- Clone:S97A-31
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Wet Ice
- Purification:Protein G purified
- Cat. No.:75930-334
- Supplier no.:200-302-W75
Neuroligin-1 is a neuronal cell surface protein belonging to the type-B carboxylesterase/lipase family. It is a necessary component in the maturation of excitatory synapses for their normal, functional development, and is necessary to the regulation of synaptic plasticity and the development of long-term memory within the adult amygdala in mammals. It is believed to participate in cell-cell-interaction through the assembly of intracellular junction by the binding of beta-neurexins, and may also be a factor in the maintenance and assembly of synaptic junctions. It is also thought to have involvement in excitatory synaptic specification. Within brain tissue, Neuroligin-1 is primarily observed in neurons and spinal cord. Anti-Neuroligin-1 is ideal for research in Neuroscience, including narcolepsy and synaptic neurotransmission: GABAergic inhibition, as well as Cell Adhesion.
Anti-Neuroligin 1 FITC Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~120kDa on specific lysates or tissues. Does not cross react with other Neuroligins. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: Neuroligin-1
Clonality: Monoclonal
Clone: S97A-31
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: