To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Name:Neuroligin-1
- Clonality:Monoclonal
- Clone:S97A-31
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Wet Ice
- Purification:Protein G purified
- Cat. No.:75930-310
- Supplier no.:200-301-W75
Specifications
About this item
Neuroligin-1 is a neuronal cell surface protein belonging to the type-B carboxylesterase/lipase family. It is a necessary component in the maturation of excitatory synapses for their normal, functional development, and is necessary to the regulation of synaptic plasticity and the development of long-term memory within the adult amygdala in mammals. It is believed to participate in cell-cell-interaction through the assembly of intracellular junction by the binding of beta-neurexins, and may also be a factor in the maintenance and assembly of synaptic junctions. It is also thought to have involvement in excitatory synaptic specification. Within brain tissue, Neuroligin-1 is primarily observed in neurons and spinal cord. Anti-Neuroligin-1 is ideal for research in Neuroscience, including narcolepsy and synaptic neurotransmission: GABAergic inhibition, as well as Cell Adhesion.
Anti-Neuroligin 1 Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~120kDa on specific lysates or tissues. Does not cross react with other Neuroligins. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: Neuroligin-1
Clonality: Monoclonal
Clone: S97A-31
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: