Specifications
- Antibody Type:Primary
- Antigen Name:Mitofusin-2
- Antigen Symbol:Mitofusin-2
- Clonality:Monoclonal
- Clone:S153-5
- Conjugation:RPE (R-Phycoerythrin)
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Wet Ice
- Purification:Protein G purified
- Cat. No.:75929-958
- Supplier no.:200-308-W71
Specifications
About this item
Mitofusin-2 (Mfn2) is a membrane protein of the mitochondria and is important in mitochondrial fusion in mammalian cells. Mfn2 activity has also been implicated in mitochondrial metabolism and the loss of Mfn2 may be the reason for certain metabolic modifications that are seen in individuals with obesity. Mfn2 has been associated with maintaining the membrane potential of the mitochondria. Mfn2 has been shown to hinder mitochondrial antiviral signaling by inhibiting production of type 1 interferons. Mfn2 is a p53-inducible target and involved in cell proliferation, apoptosis promotion and tumor suppression. Anti-Mitofusin-2 is ideal for research in Metabolic Processes and Hemostasis.
Anti-Mitofusin 2 R-PE Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~90kDa on specific lysates or tissues. Does not cross react with Mitofusin 1. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: Mitofusin-2
Clonality: Monoclonal
Clone: S153-5
Conjugation: RPE
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: