Specifications
- Antibody Type:Primary
- Antigen Symbol:ME2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75929-920
- Supplier no.:600-401-CS9
Specifications
About this item
ME2 is a homotetrameric, mitochondrial NAD-dependent malic enzyme that catalyzes the oxidative decarboxylation of malate to pyruvate (1). It is related to malic enzyme 1 (ME1), a cytoplasmic NADP-dependent enzyme that generates NADPH for fatty acid biosynthesis (2). The expression of both malic enzymes is reciprocally regulated by p53; this regulation has been shown to modulate metabolism and senescence (3). Certain single-nucleotide polymorphism haplotypes of the ME2 gene have been shown to increase the risk for idiopathic generalized epilepsy (4).
Anti-ME2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 65 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: ME2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: