- Antibody Type:Primary
- Antigen Symbol:MAP3K7
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75932-566
- Supplier no.:600-401-EZ6
TAK1 (TGF-beta activated protein kinase 1) is a widely expressed enzyme originally identified as a mitogen-activated protein kinase kinase kinase (MAP3K7). It has since been shown to mediate various intracellular signaling pathways, such as those induced by TGF-beta and members of the Toll-IL-1R (TIR) superfamily, thus acting as an intermediate in both proliferative and innate and adaptive immune responses. TAK1 is normally present in cells in a complex with TAK1-binding protein 1 (TAB1) and either TAB2 or TAB3. Association with TAB1 triggers TAK1 kinase activity by inducing TAK1 autophosphorylation, while TAB2 and possibly TAB3 may contribute to SAPK2a/p38a-mediated feedback inhibition of TAK1 activity.
Anti-TAK1 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 67 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: MAP3K7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: