Specifications
- Antibody Type:Primary
- Antigen Symbol:MAP2K2
- Clonality:Monoclonal
- Clone:19G10.F1.E2
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b kappa
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Ambient
- Purification:Protein A purified
- Cat. No.:76235-024
Specifications
About this item
MEK2 antibodies detect the MEK2 isoform. Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
Anti-MEK 2 Biotin Conjugated (MOUSE) Antibody is suitable for use in Western Blotting and ELISA. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 44 kDa.
Type: Primary
Antigen: MAP2K2
Clonality: Monoclonal
Clone: 19G10.F1.E2
Conjugation: BAC
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: