Specifications
- Antibody Type:Primary
- Antigen Symbol:MAP2K2
- Clonality:Monoclonal
- Clone:12A6.G1.G11
- Conjugation:FITC (Fluorescein Isothiocyanate)
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Ambient
- Purification:Protein A purified
- Cat. No.:76235-018
Specifications
About this item
MEK2 antibodies detect the MEK2 isoform. Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. This antibody does not react with MEK1. The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
Anti-MEK 2 FITC Conjugated (MOUSE) Antibody is suitable for use in Western Blotting and ELISA. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 44 kDa.
Type: Primary
Antigen: MAP2K2
Clonality: Monoclonal
Clone: 12A6.G1.G11
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: