Specifications
- Antibody Type:Primary
- Antigen Symbol:MAP2K2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. This product is stable at 4 °C as an undiluted liquid. For extended storage, aliquot contents and freeze at –20 °C or below. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:76262-698
Specifications
About this item
MEK2 antibodies detect the MEK2 isoform. Mitogen-activated protein kinase kinase 2, also known as MEK2 or MKK2, is an integral component of the MAP kinase cascade that regulates cell growth and differentiation. This pathway also plays a key role in synaptic plasticity in the brain. Activated MEK 2 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase. MEK1 and MEK2 are about 80% identical to each other, and nearly identical within the kinase domain. The MEK2 antibody is ideal for investigators involved in Neuroscience, Cell Signaling and Cancer Research.
Anti-MEK 2 (RABBIT) antibody is suitable for use in ELISA, Western Blotting, and IHC. Specific conditions of reactivity should be optimized by the end user. Expect a band of approximately 44 kDa.
Type: Primary
Antigen: MAP2K2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: