Order Entry
Puerto Rico
ContactUsLinkComponent
Anti-LRRTM4 Rabbit Polyclonal Antibody
Anti-LRRTM4 Rabbit Polyclonal Antibody
Catalog # 75929-756
Anti-LRRTM4 Rabbit Polyclonal Antibody
Catalog # 75929-756
Supplier Number:  600-401-CN2

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    LRRTM4
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Storage Temperature:
    Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
  • Shipping Temperature:
    Dry Ice
  • Purification:
    Affinity purified
  • Cat. No.:
    75929-756
  • Supplier no.:
    600-401-CN2

Specifications

About this item

The Leucine-rich repeat transmembrane neuronal proteins (LRRTMs) are differentially expressed in the nervous system and were recently found to instruct presynaptic and mediate postsynaptic glutamatergic differentiation, with LRRTM1 and LRRTM2 most potent at inducing presynaptic differentiation. Little is known about the function of LRRTM4; in mouse, it is expressed in the limb mesenchyme, neural tube, caudal mesoderm and in three distinct regions of the head. Later expression occurs in a subset of the developing sclerotome. Its similarity to other LRRTM family members suggests that LRRTM4 may also play a role in the development and maintenance of the vertebrate nervous system.

Anti-LRRTM4 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 67 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: LRRTM4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: