Order Entry
United States
ContactUsLinkComponent
 
Anti-Leptin Rabbit Polyclonal Antibody
Catalog # 76262-628
Anti-Leptin Rabbit Polyclonal Antibody
Catalog # 76262-628
Supplier Number:  210-401-320S
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    Leptin
  • Antigen Symbol:
    Leptin
  • Clonality:
    Polyclonal
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
  • Western Blot:
    Yes
  • Size:
    25 µL
  • Environmentally Preferable:
  • Shipping Temperature:
    Dry Ice
  • Purification:
    multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
  • Cat. No.:
    76262-628

Specifications

About this item

Anti-Leptin antibody detects leptin protein. Leptin may function as part of a signaling pathway that acts to regulate the size of the body fat depot. An increase in the level of LEP may act directly or indirectly on the CNS to inhibit food intake and/or regulate energy expenditure as part of a homeostatic mechanism to maintain constancy of the adipose mass. Obesity associated leptin is the product of the OB gene and has been identified with Type II diabetes. Anti-leptin Antibody is ideal for investigators involved in Cell Signaling, Obesity research, aging research and Signal Transduction research.

This IgG fraction of anti-Mouse Leptin was tested for use in ELISA and immunoblotting.  Reactivity in other immunoassays is unknown. The antibody will recognize recombinant and native 16 kDa leptin from mouse and human.

Type: Primary
Antigen: Leptin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: