Specifications
- Antibody Type:Primary
- Antigen Symbol:LAMA1/LAMB1
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Wet Ice
- Purification:immunoaffinity chromatography
- Cat. No.:75934-814
- Supplier no.:600401-116-0.1
Specifications
About this item
Laminin Antibody is specific to Laminin. Laminins are major proteins in the basal lamina, a protein network foundation for most cells and organs. The laminins are an important and biologically active part of the basal lamina, influencing cell differentiation, migration, adhesion as well as phenotype and survival. Anti-Laminin antibody is idea for investigators involved in cell cycle protein and extracelluar matrix research.
Anti-Laminin (RABBIT) Antibody was assayed by immunoblot and found to be reactive against Laminin at a dilution of 1:5,000 to 1:10,000. Anti-Laminin (Human) (RABBIT) Antibody was also assayed against 1.0 µg of Laminin in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) code #611-1302 and (ABTS (2,2'-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature. A working dilution of 1:4,000 to 1:8,000 of the stock concentration is suggested for this product. For immunohistochemistry on paraffin embedded tissue dilute the product 1:50 to 1:200. Specific conditions should be optimized by researcher.
Type: Primary
Antigen: LAMA1/LAMB1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human