Specifications
- Antibody Type:Primary
- Antigen Symbol:IQSEC2
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75929-214
- Supplier no.:600-401-BZ0
Specifications
About this item
ADP ribosylation factors (Arfs) are small GTP-binding proteins known for their role in vesicular transport, where they nucleate the assembly of coat protein complexes at sites of carrier vesicle formation. Similar to the related protein IQSEC1, IQSEC2 is a guanine nucleotide exchange protein for ARF1 and ARF6 that belongs to the BRAG family and contains one IQ domain, one PH domain and one SEC7 domain (1,2). IQSEC2 is a major component of the postsynaptic density (PSD) and colocalizes with PSD-95. It has been suggested to modulate the functions of the ARF proteins (1). Mutations in this gene have been implicated in nonsyndromic X-linked mental retardation (3).
Anti-IQSEC2 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 162 kDa in Western Blots of specific cell lysates and tissues.
Type: Primary
Antigen: IQSEC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: