Specifications
- Antibody Type:Primary
- Antigen Symbol:H2AFX
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Dry Ice
- Purification:Affinity purified
- Cat. No.:75928-676
- Supplier no.:600-401-H36
Specifications
About this item
Histone H2A.X (phospho S139) antibody is ideal for western blotting and ELISA. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. DNA is wrapped around histone-groups, consisting of the core histones H2A, H2B, H3 and H4. As a reaction on DNA Double-strand breaks (DSB) H2AX becomes phosphorylated on serine 139, called gamma-H2AX. ATM, ATR and PRKDCs, kinases of the PI3-family, are responsible for this phosphorylation. The modification can happen accidentally during replication fork collapse, exogenous genotoxic agents, may also occur during meiotic recombination events and immunoglobulin class switching in lymphocytes, in the response to ionizing radiation but also during controlled physiological processes such as V(D)J recombination. Mutagenesis experiments have shown that the modification is necessary for the proper formation of ionizing radiation induced foci in response to double strand breaks, but is not required for the recruitment of proteins to the site of DSBs. Gamma-H2AX is a sensitive target for looking at DSBs in cells. Dephosphorylation of Ser-140 by PP2A is required for DNA DSB repair. The role of the phosphorylated form of the histone in DNA repair is under. Anti-H2AX pS139 is ideal for researched interested in Histones, DNA Damage and Repair, and Epigenetics.
Anti-H2AX pS139 antibody has been tested for use in ELISA and by western blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 15 kDa in size corresponding to phosphorylated H2AX protein by western blotting in the appropriate stimulated tissue or cell lysate or extract.
Type: Primary
Antigen: H2AFX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: