To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antibody Type:Primary
- Antigen Symbol:GRIN2B
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:
- Western Blot:Yes
- Size:100 µL
- Environmentally Preferable:
- Modification:Phosphorylation
- Storage Temperature:Store vial at –20 °C prior to opening in undiluted aliquots. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Dilut
- Shipping Temperature:Wet Ice
- Purification:Affinity purified
- Cat. No.:75930-472
- Supplier no.:600-401-X17
Specifications
About this item
The NMDA receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimer’s, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B-subunit of the NMDA Receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Phosphorylation of Ser1166 is thought to play an essential role in memory and neuronal development. NMDA pS1166 Recepor NR2B Subunit Antibody is ideal for researchers interested in neuroscience research.
Anti-Phospho-Ser1166 NMDA Receptor NR2B Subunit Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~180 kDa NMDAR NR2B subunit phosphorylated at Ser1166. Immunolabeling of the NMDA NR2B subunit band is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide. Specific conditions for reactivity should be optimized by the end user.
Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: