Specifications
- Antibody Type:Primary
- Antigen Symbol:Alb
- Clonality:Polyclonal
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Western Blot:Yes
- Size:1 mg
- Storage Temperature:Store vial at –20 °C. Aliquot contents and freeze at –20 °C. Centrifuge product if not completely clear after standing at room temperature. Stable for several weeks at 4 °C as an undiluted liquid. Dilute only prior to immediate use.
- Shipping Temperature:Dry Ice
- Purification:immunoaffinity chromatography
- Cat. No.:75933-966
- Supplier no.:600-401-254
Specifications
About this item
Anti-Mouse Serum Albumin antibody detects Serum Albumin. Serum albumin is the main protein of plasma. It has a good binding capacity for water, Ca2+, Na+, K+, fatty acids, hormones, bilirubin and drugs. Its main function is the regulation of the colloidal osmotic pressure of blood. ALB is the major zinc transporter in plasma; it typically binds about 80% of all plasma zinc. Mouse serum antibody, BSA antibody, and other albumin antibodies are ideal for investigators involved in Cell Signaling, Neuroscience, and Signal Transduction research.
Anti-ALB antibody has been assayed against 1.0 µg of Mouse Serum Albumin in a standard ELISA using Peroxidase conjugated Affinity Purified Goat anti-Rabbit IgG [H&L] MX code #611-103-122 and TMB as a substrate for 30 minutes at room temperature.
Type: Primary
Antigen: Alb
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: