Specifications
- Antibody Type:Primary
- Antigen Symbol:A2M
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- Western Blot:Yes
- Size:100 µg
- Shipping Temperature:Ambient
- Purification:multi-step process which includes delipidation, salt fractionation and ion exchange chromatography
- Cat. No.:10799-976
- Supplier no.:200106207-0100
Specifications
About this item
Anti-alpha-2-Macroglobulin antibody is secreted into the plasma and belongs to the protease inhibitor I39 family. Anti-alpha-2-Macroglobulin inhibits all proteinases utilizing a “bait region,” or specific cleavage site, which traps the proteinase, after which a thioester bond is hydrolyzed and regulates the covalent binding of the protein to proteinase. Anti-alpha-2-Macroglobulin antibody is ideal for investigators interested in Cardiovascular , Protease Inhibitor, or Cell Biology research.
Anti-Alpha-2-Macroglobulin has been assayed against 1.0 ug of alpha-2-MACROGLOBULIN in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature. A working dilution of 1:4,000 to 1:20,000 is suggested for this product.
Type: Primary
Antigen: A2M
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity: