Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Ammonia solution 14%

Ammonia solution 14%

PE drum with steel outer, 370 lbs.
Catalog # JT0889-15
Corrosive
Irritant
Environment
Supplier #:  0889-15
Ammonia solution 14%
Catalog # JT0889-15
CAS Number:  1336-21-6
MOC:  Regulated LimitedCertificates
Ammonia solution 14%
Ammonia solution 14%

PE drum with steel outer, 370 lbs.

CAS Number:  1336-21-6
MOC:  Regulated Limited
Price
$4,741.20
Log in for availability
Case (370lb)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Synonyms:  Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water
  • CAS Number:1336-21-6
  • Formula:NH₃
  • MDL Number:MFCD00066650
  • Molecular Weight:35.05 g/mol
  • Density:0.90 g/cm³
  • UN:2672
  • ADR:8,III
  •  :Management of Change (MOC) category = RL

Documents

Specifications

Specification Test Results

  • Appearance:
    Passes Test
  • Assay (as NH₃):
    11.2 - 16.8 %

Specifications

  • Pack type:
    PE drum with steel outer
  • Size:
    370 lbs.

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.