ProXtract B™ Extraction Buffer, Rockland Immunochemicals
About this item
ProXtract B™ is ready-to-use as a working 1X solution and requires no further dilution.
- Ready-to-Use
- For protein extraction
- Gentle and has a high efficiency of protein recovery
- Compatible
- No interference
This buffer can be stored and used at room temperature. ProXtract B™ Extraction Buffer is gentle and has a high efficiency of protein recovery. It is compatible with chelating agents, reducing agents, protease inhibitors, cocktail inhibitors and chaotropic salts. There is no interference with downstream purification processes. Other applications may require detergents or other additional components.
Application Note: ProXtract B™ Buffer is suitable for protein extraction and easily scalable. Specific conditions should be optimized by the end user. See protocol.
2 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
ProXtract B™ is ready-to-use as a working 1X solution and requires no further dilution.
- Ready-to-Use
- For protein extraction
- Gentle and has a high efficiency of protein recovery
- Compatible
- No interference
This buffer can be stored and used at room temperature. ProXtract B™ Extraction Buffer is gentle and has a high efficiency of protein recovery. It is compatible with chelating agents, reducing agents, protease inhibitors, cocktail inhibitors and chaotropic salts. There is no interference with downstream purification processes. Other applications may require detergents or other additional components.
Application Note: ProXtract B™ Buffer is suitable for protein extraction and easily scalable. Specific conditions should be optimized by the end user. See protocol.
Product Family Options
Product Information
- SizeAvailabilityPrice
- RLMB0870100ProXtract B™ Extraction BufferSupplier Number: MB-087-0100Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- RLMB0870500ProXtract B™ Extraction BufferSupplier Number: MB-087-0500Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.