Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
 

Normal Saline

Normal Saline, 1 L
  76237-548
 :  MB-049-1000
Normal Saline
  76237-548
 :  
Normal Saline

Normal Saline, 1 L

 :  
 
$94.96
   
Each (1l)
 
 

This solution is commonly referred to as physiological saline or isotonic saline.

  • 0.15 M Sodium Chloride
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers
  • Tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria

Application Note: Saline solutions are available in various formulations for different purposes. Salines are also used in cell biology, molecular biology, and biochemistry experiments where ever normal salines are needed.

 

 

  • Description:
    Normal Saline
  • Size:
    1 L
  • Cat. No.:
    76237-548