Order Entry
Puerto Rico
ContactUsLinkComponent
Potassium Acetate 5.0 M, Rockland Immunochemicals
Potassium Acetate 5.0 M, Rockland Immunochemicals
Catalog # 10799-504
CAS Number:  
Potassium Acetate 5.0 M, Rockland Immunochemicals
Catalog # 10799-504
Supplier Number:  MB-042-0500
CAS Number:  

Specifications

  • Description:
    5.0 M Potassium Acetate
  • Size:
    500 mL
  • Cat. No.:
    10799-504
  • Supplier no.:
    MB-042-0500

Specifications

About this item

5.0 M Potassium Acetate (untitrated) DEPC Treated

  • 5.0M concentrated stock solution
  • Aseptically filtered through a Millipore 0.22 micron filter into clean, pre-sterilized containers

In molecular biology, potassium acetate is used to precipitate dodecyl sulfate (DS) and DS-bound proteins, allowing the removal of proteins from DNA. It is also used as a salt for the ethanol precipitation of DNA. Potassium acetate is used in mixtures applied for tissue preservation, fixation, and mummification. Potassium acetate is used as a catalyst in the production of polyurethanes.

The product was tested on trypticase soy agar for 24 hours, 48 hours and 72 hours and was found to be negative for bacteria.

Application Note: This product is a 5.0M concentrated stock solution and should be diluted appropriately with distilled, deionized water or equivalent to its final working concentration. This buffer contains untitrated 5.0 M Potassium Acetate.  Meticulously prepared using ultra pure reagents dissolved in highly polished pharmaceutical grade deionized water treated with diethyl pyrocarbonate (DEPC).