- Description:LYFO DISK Aspergillus terreus
- Cat. No.:76299-412
- ATCC no.:ATCC® 1012™*
An ideal solution for many quality control applications including presence/absence testing, microbial identification methods, antimicrobial susceptibility testing, media QC, water testing and more.
- Over 900 strains available
- Easy to use – simply re-hydrate the pellet and inoculate
- Strains are less than or equal to four passages from reference culture
- Ready-to-use format saves time and money
- Refrigerated storage is easy and economical
- Online Certificate of Analysis provides detailed strain information
- Traceability to reference cultures ensures authenticity
It doesn’t get much easier than this. Simply rehydrate LYFO DISK qualitative microorganism pellets with a diluent, such as phosphate buffer or saline, and you’re ready to inoculate your culture media. LYFO DISK, packaged in a glass vial containing 6 pellets of a single strain, offers a ton of flexibility.