Human HLA-DRB1*01:01 Tetramer, PE (Phycoerythrin)
Catalog # 103501-044
Supplier: Avantor
CAS Number:
Specifications
- Conjugation:PE (Phycoerythrin)
- Species:Human
- Size:50 tests
- Protein/Peptide Name:HLA-DRB1*01:01 Tetramer
- Cat. No.:103501-044
Specifications
About this item
MHC tetramers for detecting antigen specific t-cell populations.