ContactUsLinkComponent
HeLa Cell Nuclear Extract Nocodozole Stimulated, Rockland Immunochemicals
  75928-454
 :  
HeLa Cell Nuclear Extract Nocodozole Stimulated, Rockland Immunochemicals
  75928-454
 :  W09-001-A84
 :  
  • Description:
    HeLa Cell Nuclear Extract Nocodozole Stimulated
  • Size:
    200 µg
  • Shipping Conditions:
    Dry Ice
  • Cat. No.:
    75928-454
  • Supplier no.:
    W09-001-A84
HeLa Cell Nuclear Extract Nocodozole Stimulated
  • Ready-to-use lysates
  • positive controls
  • treated lysated
  • nuclear lysates
Ready-to-use nuclear extracts produced by Rockland Immunochemicals are derived from cell lines or tissues using highly refined extraction protocols to ensure exceptionally high quality, protein integrity and lot-to-lot reproducibility. All extracts are tested by SDS-PAGE using 4-20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.
Ready-to-use nuclear extracts are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Nuclear extracts are supplied in denaturing buffer without dissociating agents. Heat nuclear extract to 95° C for 5 minutes and rapidly cool. If dissociating conditions are desired add reducing agent prior to heating. The recommended loading volume per lane is 10-30 l depending on the size format of your gel.