Order Entry
Puerto Rico
ContactUsLinkComponent
 
HeLa Whole Cell Lysate Nocodozole Stimulated, Rockland Immunochemicals
Catalog # 75934-382
CAS Number:  
HeLa Whole Cell Lysate Nocodozole Stimulated, Rockland Immunochemicals
Catalog # 75934-382
Supplier Number:  W09-001-A81
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    HeLa Whole Cell Lysate Nocodozole Stimulated
  • Size:
    500 µg
  • Shipping Conditions:
    Dry Ice
  • Cat. No.:
    75934-382
  • Supplier no.:
    W09-001-A81

Specifications

About this item

HeLa Whole Cell Lysate Nocodozole Stimulated

  • Ready-to-use lysates
  • positive controls
  • treated lysated

Ready-to-use whole cell lysates produced by Rockland Immunochemicals are derived from cell lines or tissues using highly refined extraction protocols to ensure exceptionally high quality, protein integrity and lot-to-lot reproducibility.  All extracts are tested by SDS-PAGE using 4-20% gradient gels and immunoblot analysis using antibodies to key cell signaling components to confirm the presence of both high molecular weight and low molecular weight proteins.

Ready-to-use lysates are especially prepared as positive controls for separation by SDS-PAGE and subsequent western blot analysis. Lysates are prepared in denaturing buffer WITHOUT dissociating agents (i.e. no 2-mercaptoethanol or dithiothreitol has been added). Heat lysate to 95° C for 5 minutes and rapidly cool.  If dissociating conditions are desired, add reducing agent prior to heating.  The recommended loading volume per lane is 10-20 µl depending on the size format of your gel.