Specifications
- Description:Goat Blood In Sodium Citrate
- Size:50 mL
- Environmentally Preferable:
- Shipping Temperature:Wet Ice
- Cat. No.:10801-648
- Supplier no.:R204-0050
Specifications
About this item
Goat Blood In Sodium Citrate
- aseptically collected and prepared to order
- blood draw in Anticoagulant
Citrated goat blood is aseptically collected and prepared to order from our colony of animals. These cell suspensions are useful for the titration of complement, adsorption procedures, testing for agglutinins/HA assays, and for the preparation of stroma as particulate reagents. Goat red blood cells are perishable and are collected and processed upon receipt of your order.
Washed pooled cells preparations, complement titration, adsorption procedures, HA assays and for the preparation of stroma as particulate reagents.