Sheep Peroxidase Anti-Peroxidase (PAP) Soluble Complex, Rockland Immunochemicals
75931-862
:
- Conjugation:HRP (Horseradish Peroxidase)
- Description:Sheep Peroxidase Anti-Peroxidase (PAP) Soluble Complex
- Size:25 mg
- Shipping Temperature:Ambient
- Cat. No.:75931-862
- Supplier no.:P500-0025
Sheep Peroxidase Anti-Peroxidase (PAP) Soluble Complex
- peroxidase-anti-peroxidase
- peroxidase enzymatic labeling
- amplification of signal, indirectly detecting the amount of primary antibody
The peroxidase-antiperoxidase (PAP) approach utilizes a complex of enzyme and antibody to facilitate amplification of signal, indirectly detecting the amount of primary antibody used in an assay. Generated in Sheep, the PAP complex can be utilized in any assay that utilizes a sheep primary antibody. With the addition of a secondary antibody that recognizes sheep, the reporter PAP complex allows for amplification of the luminescent signal (as three molecules of enzyme are bound to every two immunoglobulins).
Suitable for use in peroxidase enzymatic labeling experiments.