ContactUsLinkComponent
SDS-PAGE Destain Solution, Rockland Immunochemicals
SDS-PAGE Destain Solution, Rockland Immunochemicals
Catalog # 75931-768
CAS Number:  
SDS-PAGE Destain Solution, Rockland Immunochemicals
Catalog # 75931-768
Supplier Number:  MB-022-1000
CAS Number:  
Specifications
  • Size:
    1 L
  • Cat. No.:
    75931-768
  • Supplier no.:
    MB-022-1000
Specifications
About this item
SDS-PAGE Destain Solution
  • ready-to-use as a working 1X solution
  • in-gel visualization of Coomassie Blue stained total protein
Destain Solution is a uniquely formulated, ready-to-use reagent specifically designed for in-gel visualization of Coomassie Blue stained total protein in polyacrylamide gels. The proprietary formulation of the solution ensures high detection sensitivity and low backgrounds.
This product is ready-to-use as a working 1X solution and requires no further dilution. This solution contains alcohol and acetic acid in a proprietary combination to enhance sensitivity/noise ratio.