Order Entry
Puerto Rico
ContactUsLinkComponent
Human APOE ELISA Kit, Rockland Immunochemicals
Human APOE ELISA Kit, Rockland Immunochemicals
Catalog # 76236-272
CAS Number:  
Human APOE ELISA Kit, Rockland Immunochemicals
Catalog # 76236-272
Supplier Number:  KOA0878
CAS Number:  

Specifications

  • Primary antibody reactivity:
    Human
  • Description:
    Human APOE ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-272

Specifications

About this item

Human APOE AccuSignal ELISA Kit

  • Natural human APOE.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: K19-H317

Apolipoprotein E (APOE) is a class of apolipoprotein found in the chylomicron and Intermediate-density lipoprotein (IDLs) that is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. In peripheral tissues, APOE is primarily produced by the liver and macrophages, and mediates cholesterol metabolism in an isoform- dependent manner. In the central nervous system, APOE is mainly produced by astrocytes, and transports cholesterol to neurons via APOE receptors, which are members of the low density lipoprotein receptor gene family. This protein is involved in Alzheimer’s disease and cardiovascular disease.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 200ng/ml, 100ng/ml, 50ng/ml, 25ng/ml, 12.5ng/ml, 6.25ng/ml, 3.12ng/ml human APOE standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum, plasma(heparin, EDTA), saliva, milk or urine to each empty well. It is recommended that each human APOE standard solution and each sample be measured in duplicate.