- Primary antibody reactivity:Human
- Description:Human IDS ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-268
Human IDS AccuSignal ELISA Kit
- Natural and recombinant human IDS.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: S26-P550
Iduronate 2-sulfatase (IDS) is a sulfatase enzyme associated with Hunter syndrome. It encodes a member of the sulfatase family of proteins. Iduronate 2-sulfatase is involved in the lysosomal degradation of the glycosaminoglycans heparan sulfate and dermatan sulfate. The encoded preproprotein is proteolytically processed to generate two polypeptide chains. Mutations in this gene are associated with the X-linked lysosomal storage disease mucopolysaccharidosis type II, also known as Hunter syndrome. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human IDS standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human IDS standard solution and each sample be measured in duplicate.