Specifications
- Assay Type (ELISA with LOV):Sandwich
- Description:Rat CD44 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-242
Specifications
About this item
Rat CD44 AccuSignal ELISA Kit
- Natural and recombinant rat CD44.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: Q22-E271
CD44 is an integral cell membrane glycoprotein with a postulated role in matrix adhesion lymphocyte activation and lymph node homing. It is contains 19 exons spanning 50 kb of genomic DNA. In humans, the CD44 antigen is encoded by the CD44 gene on Chromosome 11. The protein encoded by this gene is a cell-surface glycoprotein involved in cell-cell interactions, cell adhesion and migration. It is a receptor for hyaluronic acid (HA) and can also interact with other ligands, such as osteopontin, collagens, and matrix metalloproteinases (MMPs). Transcripts for this gene undergo complex alternative splicing that results in many functionally distinct isoforms, however, the full length nature of some of these variants has not been determined. Alternative splicing is the basis for the structural and functional diversity of this protein, and may be related to tumor metastasis.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 20,000pg/ml, 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml rat CD44 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each rat CD44 standard solution and each sample be measured in duplicate.