- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse CD5L ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-240
Mouse CD5L AccuSignal ELISA Kit
- Natural and recombinant mouse CD5L.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: E22-V352
CD5 antigen-like, also known as Sp alpha and AIM, is a protein that in humans is encoded by the CD5L gene. It is mapped to 1q21-q23 by fluorescence in situ hybridization. It is found that Aim expression is induced in mouse macrophages in response to loading with highly oxidized low density lipoprotein (oxLDL), and that Aim is expressed in foam cells within atherosclerotic lesions. Both the expression of Aim in lesions and its induction by oxLDL require Lxr /Rxr heterodimers. Aim-null macrophages are highly susceptible to oxLDL-induced apoptosis in vitro and undergo accelerated apoptosis in atherosclerotic lesions in vivo. Double knockout of Aim and Ldlr reduce atherosclerotic lesions. Therefore, it is concluded that AIM expression protects macrophages from apoptosis within atherosclerotic lesions, promoting early lesion development.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 313pg/ml, 156pg/ml mouse CD5L standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. It is recommended that each mouse CD5L standard solution and each sample be measured in duplicate.