Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse FGF21 ELISA Kit, Rockland Immunochemicals
Mouse FGF21 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-228
CAS Number:  
Mouse FGF21 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-228
Supplier Number:  KOA0836
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse FGF21 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-228

Specifications

About this item

Mouse FGF21 AccuSignal ELISA Kit

  • Natural and recombinant mouse FGF21.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: A29-S210

Fibroblast growth factor 21 is a protein that in humans is encoded by the FGF21 gene. By genomic sequence analysis, Nishimura et al. (2000) identified the FGF21 gene within the 5-prime flanking region of the FUT1 gene on chromosome 19. The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. FGF21 stimulates glucose uptake in adipocytes but not in other cell types. This effect is additive to the activity of insulin. FGF21 treatment of adipocytes is associated with phosphorylation of FRS2, a protein linking FGF receptors to the Ras/MAP kinase pathway. FGF21 injection in ob/ob mice results in an increase in Glut1 in adipose tissue. FGF21 also protects animals from diet-induced obesity when overexpressed in transgenic mice and lowers blood glucose and triglyceride levels when administered to diabetic rodents.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 2000pg/ml,1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml mouse FGF21 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma (heparin, EDTA) to each empty well. It is recommended that each mouse FGF21 standard solution and each sample be measured in duplicate.