- Primary antibody reactivity:Human
- Description:Human ADAMTS4 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-218
Human ADAMTS4 AccuSignal ELISA Kit
- Natural and recombinant human ADAMTS4.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: CHO; Immunogen sequence: F213-C685
ADAMTS4, A disintegrin and metalloproteinase with thrombospondin motifs 4 is an enzyme that in humans is encoded by the ADAMTS4 gene. ADAMTS4 is a member of the large ADAMTS family of zinc-dependent proteases. The human ADAMTS4 gene is mapped to chromosome 1 by somatic cell hybrid analysis. The enzyme encoded by this gene lacks a C-terminal TS motif. It is responsible for the degradation of aggrecan, a major proteoglycan of cartilage, and brevican, a brain-specific extracellular matrix protein. The cleavage of aggrecan and brevican suggests key roles of this enzyme in arthritic disease and in the central nervous system, potentially, in the progression of glioma.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 40,000pg/ml, 20,000pg/ml, 10,000pg/ml, 5,000pg/ml, 2,500pg/ml, 1,250pg/ml, 625pg/ml human ADAMTS4 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each human ADAMTS4 standard solution and each sample be measured in duplicate.