Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Mouse Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-212
CAS Number:  
Mouse Tissue factor - F3 ELISA Kit, Rockland Immunochemicals
Catalog # 76236-212
Supplier Number:  KOA0828
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse Tissue factor - F3 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76236-212

Specifications

About this item

Mouse Tissue factor - F3 AccuSignal ELISA Kit

  • Natural and recombinant mouse Tissue factor.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: NSO; Immunogen sequence: A29-E251

Tissue factor, also called platelet tissue factor, factor III, thrombokinase, or CD142 is a protein present in sub endothelial tissue and leukocytes necessary for the initiation of thrombin formation from the zymogene prothrombin. Tissue factor gene was mapped to 1pter-p21 by study of somatic cell hybrids with a species-specific sensitive chromogenic assay. This gene encodes coagulation factor III which is a cell surface glycoprotein. This factor enables cells to initiate the blood coagulation cascades, and it functions as the high-affinity receptor for the coagulation factor VII.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse Tissue factor standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA, citrate) or cell lysates to each empty well.. It is recommended that each mouse Tissue factor standard solution and each sample be measured in duplicate.