Order Entry
Puerto Rico
ContactUsLinkComponent
Mouse FGF23 ELISA Kit, Rockland Immunochemicals
Mouse FGF23 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-522
CAS Number:  
Mouse FGF23 ELISA Kit, Rockland Immunochemicals
Catalog # 76237-522
Supplier Number:  KOA0823
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Primary antibody reactivity:
    Mouse
  • Description:
    Mouse FGF23 ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-522

Specifications

About this item

Mouse FGF23 AccuSignal ELISA Kit

  • Natural and recombinant mouse FGF23.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: Y25-V251

Fibroblast growth factor 23 or FGF23 is a protein that in humans is encoded by the FGF23 gene. It is a member of the fibroblast growth factor (FGF) family which is responsible for phosphate metabolism. The main function of FGF23 seems to be regulation of phosphate concentration in plasma. FGF23 is secreted by Osteocytes in response to elevated Calcitriol. And it acts on the kidneys, where it decreases the expression of NPT2, a sodium-phosphate cotransporter in the proximal tubule. Thus, FGF23 decreases the reabsorption and increases excretion of phosphate. Also, FGF23 may suppress 1-alpha-hydroxylase, reducing its ability to activate vitamin D and subsequently impairing calcium absorption.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 1000pg/ml, 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.2pg/ml, 15.6pg/ml mouse FGF23 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse FGF23 standard solution and each sample be measured in duplicate.