Specifications
- Primary antibody reactivity:Human
- Description:Human TFPI2 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-518
Specifications
About this item
Human TFPI2 AccuSignal ELISA Kit
- Natural and recombinant human TFPI2.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: D23-F235
TFPI2, also called Tissue factor pathway inhibitor 2, is a protein that in humans is encoded by the TFPI2 gene. It is mapped to 7q21.3. This gene encodes a member of the Kunitz-type serine proteinase inhibitor family. The protein can inhibit a variety of serine proteases including factor VIIa/tissue factor, factor Xa, plasmin, trypsin, chymotryspin and plasma kallikrein. This gene has been identified as a tumor suppressor gene in several types of cancer. Qualitative PCR and microarray analyses showed that expression of TFPI2 was inversely correlated with that of MIR616 in prostate cancer cell lines, primary tumors of various grades, and normal tissue.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml human TFPI2 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of human cell culture supernates or serum to each empty well. It is recommended that each human TFPI2 standard solution and each sample be measured in duplicate.