- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse ANGPTL3 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-508
Mouse ANGPTL3 AccuSignal ELISA Kit
- Natural and recombinant mouse ANGPTL3.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: sf21; Immunogen sequence: S17-T455
ANGPTL3, also known as Angiopoietin-like 3 or HYPLl, is a protein that in humans is encoded by the ANGPTL3 gene. It is mapped to 1p31.3. The protein encoded by this gene is a member of the angiopoietin-like family of secreted factors. It is expressed predominantly in the liver, and has the characteristic structure of angiopoietins, consisting of a signal peptide, N-terminal coiled-coil domain, and the C-terminal fibrinogen (FBN)-like domain. ANGPTL3 also acts as dual inhibitor of lipoprotein lipase (LPL) and endothelial lipase (EL), and increases plasma triglyceride and HDL cholesterol in rodents. It can inhibit endothelial lipase to catalyze HDL-phospholipid and increase HDL-PL levels.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse ANGPTL3 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse ANGPTL3 standard solution and each sample be measured in duplicate.