Order Entry
Puerto Rico
ContactUsLinkComponent
Rat Adiponectin ELISA Kit, Rockland Immunochemicals
Rat Adiponectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-622
CAS Number:  
Rat Adiponectin ELISA Kit, Rockland Immunochemicals
Catalog # 76237-622
Supplier Number:  KOA0797
CAS Number:  

Specifications

  • Assay Type (ELISA with LOV):
    Sandwich
  • Description:
    Rat Adiponectin ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-622

Specifications

About this item

Rat Adiponectin AccuSignal ELISA Kit

  • Natural and recombinant rat Adiponectin.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: I19-N244

Adiponectin(ADPN) is a hormone secreted by adipocytes that regulates energy homeostasis and glucose and lipid metabolism. Adiponectin is a new member of the family of soluble defense collagens, in hematopoiesis and immune responses. It is an important negative regulator in hematopoiesis and immune systems and raise the possibility that it may be involved in ending inflammatory responses through its inhibitory functions. Adiponectin is mapped to 3q27 and can protect the organism from systemic inflammation by promoting the clearance of early apoptotic cells by macrophages through a receptor-dependent pathway involving calreticulin.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml rat Adiponectin standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum, plasma(heparin, EDTA), tissue homogenates or urine to each empty well. It is recommended that each rat Adiponectin standard solution and each sample be measured in duplicate.