Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse CD22 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76236-166
Specifications
About this item
Mouse CD22 AccuSignal ELISA Kit
- Natural and recombinant mouse CD22.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: S22-R702
CD22 or cluster of differentiation-22, is a molecule belonging to the SIGLEC family of lectins. This gene is mapped to 19q13.2. It is found on the surface of mature B cells and to a lesser extent on some immature B cells. Generally speaking, CD22 is a regulatory molecule that prevents the overactivation of the immune system and the development of autoimmune diseases. It is a negative regulator of antigen receptor signaling whose onset of expression at the mature B cell stage may serve to raise the antigen concentration threshold required for B cell triggering. CD22 functions as an inhibitory receptor for B cell receptor (BCR) signalling. This gene can downmodulate signaling through the IgM and IgD B-cell receptors(BCRs), but not through the IgG BCR, because the IgG cytoplasmic tail prevents CD22 phosphorylation and actually enhances IgG-BCR signaling.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse CD22 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernatants, serum or plasma(heparin, EDTA) to each empty well. It is recommended that each mouse CD22 standard solution and each sample be measured in duplicate.