Order Entry
Puerto Rico
ContactUsLinkComponent
 
Rat BTC ELISA Kit, Rockland Immunochemicals
  76237-482
 :  
Rat BTC ELISA Kit, Rockland Immunochemicals
  76237-482
 :  KOA0771
 :  

 

  • Assay Type (ELISA with LOV):
    Sandwich
  • Description:
    Rat BTC ELISA Kit
  • Size:
    1 Kit
  • Shipping Temperature:
    Wet Ice
  • Cat. No.:
    76237-482

 

 

Rat BTC AccuSignal ELISA Kit

  • Natural and recombinant rat BTC.
  • There is no detectable cross-reactivity with other relevant proteins.
  • Expression system for standard: E.coli; Immunogen sequence: D32-Y111

Betacellulin, also known as BTC is a protein that in humans is encoded by the BTC gene. This gene is mapped to 4q13.3. The protein encoded by this gene is a member of the EGF family of growth factors. It is synthesized primarily as a transmembrane precursor, which is then processed to mature molecule by proteolytic events. This protein is a ligand for the EGF receptor. It is the growth factor that binds to EGFR, ERBB4 and other EGF receptor family members. This protein is a potent mitogen for retinal pigment epithelial cells and vascular smooth muscle cells.

Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 500pg/ml, 250pg/ml, 125pg/ml, 62.5pg/ml, 31.3pg/ml, 15.6pg/ml, 7.8pg/ml rat BTC standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of rat cell culture supernates, serum, plasma(heparin, EDTA), tissue homogenates or urine to each empty well. It is recommended that each rat BTC standard solution and each sample be measured in duplicate.