- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse DMP-1 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76237-472
Mouse DMP-1 AccuSignal ELISA Kit
- Natural and recombinant mouse DMP-1.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: NSO; Immunogen sequence: L17-Y503
Dentin matrix acidic phosphorprotein 1 is a protein that in humans is encoded by the DMP1 gene. It belongs to the small integrin-binding ligand N-linked glycoprotein(SIBLING) family of secreted phosphoproteins. This gene is mapped to 4q22.1. This protein, which is critical for proper mineralization of bone and dentin, is present in diverse cells of bone and tooth tissues. In undifferentiated osteoblasts, DMP-1 is primarily a nuclear protein that regulates the expression of osteoblast-specific genes. During osteoblast maturation, the protein becomes phosphorylated and is exported to the extracellular matrix, where it orchestrates mineralized matrix formation. Mutations in the gene are known to cause autosomal recessive hypophosphatemia, a disease that manifests as rickets and osteomalacia. The gene structure is conserved in mammals.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 10,000pg/ml, 5000pg/ml, 2500pg/ml, 1250pg/ml, 625pg/ml, 312pg/ml, 156pg/ml mouse DMP-1 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum, plasma(heparin, EDTA) or tissue homogenates to each empty well. It is recommended that each mouse DMP-1 standard solution and each sample be measured in duplicate.