Specifications
- Assay Type (ELISA with LOV):Sandwich
- Primary antibody reactivity:Mouse
- Description:Mouse Galectin-7 ELISA Kit
- Size:1 Kit
- Shipping Temperature:Wet Ice
- Cat. No.:76235-974
Specifications
About this item
Mouse Galectin-7 AccuSignal ELISA Kit
- Natural and recombinant mouse Galectin-7.
- There is no detectable cross-reactivity with other relevant proteins.
- Expression system for standard: E.coli; Immunogen sequence: S2-F136
Galectin-7, also known as GAL7, is a protein that in humans is encoded by the LGALS7 gene. This gene is mapped to 19q13.2. Galectins belong to a family of related beta-galactoside-binding lectins, also referred to as S-type or S-Lac lectins. Members of this family have been implicated in a variety of functions, including growth regulation, cell adhesion, migration, neoplastic transformation, and immune responses. Differential and in situ hybridizations indicate that this lectin is specifically expressed in keratinocytes. It has been found that expression of Galectin-7 was markedly upregulated in corneal epithelium after injury and that exogenous galectin-7 stimulated reepithelialization of corneal wounds.
Useful in Sandwich ELISA for Quantitative Detection of Antigen. Aliquot 0.1ml per well of the 6000pg/ml, 3000pg/ml, 1500pg/ml, 750pg/ml, 419pg/ml, 187.5pg/ml, 93.8pg/ml mouse Galectin-7 standard solutions into the precoated 96-well plate. Add 0.1ml of the sample diluent buffer into the control well (Zero well). Add 0.1ml of each properly diluted sample of mouse cell culture supernates, serum or plasma(heparin, EDTA) to each empty well. We recommend that each mouse Galectin-7 standard solution and each sample is measured in duplicate.